2,4,6-Tri-tert-butylnitrobenzene ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T1821-25G
T1821-5G
TCIAT1821-25GEA
262
EUR
TCIAT1821-25G
TCIAT1821-5G
2,4,6-Tri-tert-butylnitrobenzene ≥95.0% (by GC)
2,4,6-Tri-tert-butylnitrobenzene
Formule:
C₁₈H₂₉NO₂ Dichtheid: 1,05 g/cm³ (19.85 °C) |
MDL Nummer:
MFCD00008818 CAS nummer: 4074-25-3 EINECS: 223-789-5 |
|
Specification Test Results
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 205.0 to 208.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...