Triethyl orthopropionate ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

O0067-25ML O0067-500ML
TCIAO0067-25MLEA 17.6 EUR
TCIAO0067-25ML TCIAO0067-500ML
Triethyl orthopropionate ≥96.0% (by GC)
Triethyl orthopropionate
Formule: C₉H₂₀O₃
Kookpunt: 158°C
Smeltpunt: < 0 °C
Dichtheid: 0,8876 g/cm³ (20 °C)
Vlampunt: 47°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009226
CAS nummer: 115-80-0
EINECS: 204-108-0
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c4841a5da826ee723e5f127c01218bf8aaafa8d&vid=5c4841a5da826ee723e5f127c01218bf8aaafa8d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR