Allyl ethyl ether ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Allyl Ethyl Ether
A1033-25ML A1033-5ML
TCIAA1033-25MLEA 71.5 EUR
TCIAA1033-25ML TCIAA1033-5ML
Allyl ethyl ether ≥95.0% (by GC)
Allyl ethyl ether
Formule: C₅H₁₀O
Kookpunt: 67°C
Dichtheid: 0,7559 g/cm³ (20 °C)
Vlampunt: -20°C
MDL Nummer: MFCD00009255
CAS nummer: 557-31-3
EINECS: 209-169-7
UN: 2335
ADR: 3,II
Merck Index: 14,00293
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f994ba93bb6c24ead634c53eaca1720f073f3428&vid=f994ba93bb6c24ead634c53eaca1720f073f3428&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %