Glutaric anhydride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Dihydro-2H-pyran-2,6(3H)-dione, Dihydro-2H-pyran-2,6(3H)-dione
G0071-25G
G0071-500G
G0071-100G
TCIAG0071-25GEA
18.7
EUR
TCIAG0071-25G
TCIAG0071-500G
TCIAG0071-100G
Glutaric anhydride ≥98.0% (by titrimetric analysis)
Glutaric anhydride
Formule:
C₅H₆O₃ Kookpunt: 287 °C (1013 hPa) Smeltpunt: 52…55 °C Dichtheid: 1,411 g/cm³ (20 °C) Vlampunt: 174 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006679 CAS nummer: 108-55-4 EINECS: 203-593-6 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (Morpholine Method) | min. 98.0 % |
Melting point | 55.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...