Cinnamyl alcohol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
C0362-25G C0362-500G
TCIAC0362-25GEA 19.8 EUR
TCIAC0362-25G TCIAC0362-500G
Cinnamyl alcohol ≥97.0% (by GC)
Cinnamyl alcohol
Formule: C₉H₁₀O
MW: 134.18 g/mol
Kookpunt: 250°C
Vlampunt: 126
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002921
CAS nummer: 104-54-1
EINECS: 203-212-3
Merck Index: 14,02302
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f15329783a0eaf0f84b1449ef2f576197b96de4&vid=3f15329783a0eaf0f84b1449ef2f576197b96de4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %