(±)-3-Butyne-2-ol ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B1001-25ML B1001-5ML
TCIAB1001-25MLEA 137 EUR
TCIAB1001-25ML TCIAB1001-5ML
(±)-3-Butyne-2-ol ≥97.0% (by GC)
(±)-3-Butyne-2-ol
Formule: C₄H₆O
Kookpunt: 110°C
Smeltpunt: –12 °C
Dichtheid: 0,895 g/cm³ (20 °C)
Vlampunt: 34°C
MDL Nummer: MFCD00004541
CAS nummer: 2028-63-9
EINECS: 217-978-1
UN: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d3538a4cc333e7160ffa74384d22f84bd675a3b&vid=6d3538a4cc333e7160ffa74384d22f84bd675a3b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %