1,1,3,3-Tetramethyldisilazane ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0833-25ML T0833-5ML
TCIAT0833-25MLEA 84.7 EUR
TCIAT0833-25ML TCIAT0833-5ML
1,1,3,3-Tetramethyldisilazane ≥97.0% (by GC)
1,1,3,3-Tetramethyldisilazane
Formule: C₄H₁₅NSi₂
Kookpunt: 100°C
Dichtheid: 0,766 g/cm³ (20 °C)
Vlampunt: -8°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00025626
CAS nummer: 15933-59-2
EINECS: 240-072-2
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dca2066601829ec75a176fccd03bc24969daa0a0&vid=dca2066601829ec75a176fccd03bc24969daa0a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR