Azulene ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Bicyclo[5.3.0]decapentaene
A0634-1G
A0634-100MG
A0634-5G
TCIAA0634-1GEA
304
EUR
TCIAA0634-1G
TCIAA0634-100MG
TCIAA0634-5G
Azulene ≥99.0% (by GC)
Azulene
Formule:
C₁₀H₈ MW: 128.17 g/mol Kookpunt: 242 |
MDL Nummer:
MFCD00003810 CAS nummer: 275-51-4 EINECS: 205-993-6 Merck Index: 14,00926 |
|
Specification Test Results
Appearance | Orange to Brown to Dark purple powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 99.0 to 103.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...