1-Ethylpiperazine 98,0 GC, T
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N-Ethylpiperazine
E0355-25ML
E0355-500ML
E0355-100ML
TCIAE0355-25MLEA
17.6
EUR
TCIAE0355-25ML
TCIAE0355-500ML
TCIAE0355-100ML
1-Ethylpiperazine 98,0 GC, T
1-Ethylpiperazine
Formule:
C₆H₁₄N₂ MW: 114.19 g/mol Kookpunt: 158°C Smeltpunt: -60 Vlampunt: 43°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00059912 CAS nummer: 5308-25-8 EINECS: 226-166-6 UN: 2924 ADR: 3 III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...