Pyridazine ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P0616-25G P0616-5G
TCIAP0616-25GEA 156 EUR
TCIAP0616-25G TCIAP0616-5G
Pyridazine ≥99.0% (by GC)
Pyridazine
Formule: C₄H₄N₂
Kookpunt: 208°C
Smeltpunt: -8°C
Dichtheid: 1,103 g/cm³ (20 °C)
Vlampunt: 85°C
MDL Nummer: MFCD00006463
CAS nummer: 289-80-5
EINECS: 206-025-5
Merck Index: 14,07969
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32cd5774ff53cb491f032d25b373ac4ef38b4929&vid=32cd5774ff53cb491f032d25b373ac4ef38b4929&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 99.0 %