3-Methylpyridazine ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
M0872-25ML
M0872-5ML
TCIAM0872-25MLEA
728
EUR
TCIAM0872-25ML
TCIAM0872-5ML
3-Methylpyridazine ≥97.0% (by GC)
3-Methylpyridazine
Formule:
C₅H₆N₂ Kookpunt: 215°C Dichtheid: 1,0491 g/cm³ (20 °C) Vlampunt: 87°C |
MDL Nummer:
MFCD00006469 CAS nummer: 1632-76-4 EINECS: 216-640-0 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 97.0 % |