4-Methylpyrimidine ≥96.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

M0356-25G M0356-5G
TCIAM0356-25GEA 149 EUR
TCIAM0356-25G TCIAM0356-5G
4-Methylpyrimidine ≥96.0% (by titrimetric analysis)
4-Methylpyrimidine
Formule: C₅H₆N₂
Kookpunt: 142°C
Smeltpunt: 32 °C
Dichtheid: 1,02 g/cm³ (20 °C)
Vlampunt: 40°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006115
CAS nummer: 3438-46-8
EINECS: 222-344-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ebf137eecd5954cd0baf966fcb079a7ccac0ca4&vid=6ebf137eecd5954cd0baf966fcb079a7ccac0ca4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR