4-Bromoindole ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Bromo-1H-indole
B1797-1G B1797-5G
TCIAB1797-1GEA 28.6 EUR
TCIAB1797-1G TCIAB1797-5G
4-Bromoindole ≥97.0% (by GC)
4-Bromoindole
Formule: C₈H₆BrN
MW: 196.05 g/mol
Kookpunt: 96°C
MDL Nummer: MFCD00671502
CAS nummer: 52488-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f20ec4fcca7eb69689ba73311ca02e1f4eb69f5f&vid=f20ec4fcca7eb69689ba73311ca02e1f4eb69f5f&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %