4-Bromo-3-nitrobenzotrifluoride ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-Bromo-3-nitrobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-3-nitrotoluene, 1-Bromo-2-nitro-4-(trifluoromethyl)benzene
B3074-25G B3074-5G
TCIAB3074-25GEA 142 EUR
TCIAB3074-25G TCIAB3074-5G
4-Bromo-3-nitrobenzotrifluoride ≥98.0% (by GC)
4-Bromo-3-nitrobenzotrifluoride
Formule: C₇H₃BrF₃NO₂
Kookpunt: 105°C
Dichtheid: 1,76 g/cm³ (20 °C)
MDL Nummer: MFCD00014685
CAS nummer: 349-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44c6025e74e98ddff0e50eefcdb071274562ad7b&vid=44c6025e74e98ddff0e50eefcdb071274562ad7b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %