10-Bromo-1-decanol ≥85.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 10-Bromodecanol

B1730-25G B1730-5G
TCIAB1730-25GEA 134 EUR
TCIAB1730-25G TCIAB1730-5G
10-Bromo-1-decanol ≥85.0% (by GC)
10-Bromo-1-decanol
Formule: HO(CH₂)₉CH₂Br
Kookpunt: 169°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00041681
CAS nummer: 53463-68-6
EINECS: 258-572-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bc3f508b0a2a6a23e613f7da8215023f77be34c&vid=2bc3f508b0a2a6a23e613f7da8215023f77be34c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR