Dimethyldichloorsilaan ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dichloro(dimethyl)silane, Dichlorodimethylsilane
D0358-25ML D0358-100ML
TCIAD0358-25MLEA 17.6 EUR
TCIAD0358-25ML TCIAD0358-100ML
Dimethyldichloorsilaan ≥98.0% (by GC)
Dimethyldichloorsilaan
Formule: C₂H₆Cl₂Si
MW: 129.06 g/mol
Kookpunt: 70°C
Smeltpunt: -76
Vlampunt: –7°C
MDL Nummer: MFCD00000491
CAS nummer: 75-78-5
EINECS: 200-901-0
UN: 1162
Bereik: 01-2119437250-51
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35035e0b371238b307a42c4b443a96c981e877a1&vid=35035e0b371238b307a42c4b443a96c981e877a1&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %