3'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A0104-25G
A0104-500G
TCIAA0104-25GEA
26.4
EUR
TCIAA0104-25G
TCIAA0104-500G
3'-Hydroxyacetophenone ≥98.0% (by GC, titration analysis)
3'-Hydroxyacetophenone
Formule:
C₈H₈O₂ Kookpunt: 153°C Smeltpunt: 86…88 °C Dichtheid: 1,1 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002298 CAS nummer: 121-71-1 EINECS: 204-494-0 |
|
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...