9,10-Anthraquinone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A0502-25G A0502-500G
TCIAA0502-25GEA 17.6 EUR
TCIAA0502-25G TCIAA0502-500G
9,10-Anthraquinone ≥98.0% (by GC)
9,10-Anthraquinone
Formule: C₁₄H₈O₂
Kookpunt: 380°C
Smeltpunt: 284°C
Dichtheid: 1,419 to 1.438 g/cm³ (20 °C)
Vlampunt: 185 °C
MDL Nummer: MFCD00001188
CAS nummer: 84-65-1
EINECS: 201-549-0
UN: 3077
ADR: 9,III
Merck Index: 14,00687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b08a06e493615c9382a6b8877e1de05e9c2e243&vid=6b08a06e493615c9382a6b8877e1de05e9c2e243&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %