D-(-)-Histidine ≥99.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH

H0998-25G H0998-5G
TCIAH0998-25GEA 130 EUR
TCIAH0998-25G TCIAH0998-5G
D-(-)-Histidine ≥99.0% (by HPLC, titration analysis)
D-(-)-Histidine
Formule: C₆H₉N₃O₂
MW: 155.16 g/mol
Smeltpunt: 287
MDL Nummer: MFCD00065963
CAS nummer: 351-50-8
EINECS: 206-513-8
Merck Index: 14,04720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36e8d487ca6ab7bcff8c9a8e9aba0e7178dd4bb2&vid=36e8d487ca6ab7bcff8c9a8e9aba0e7178dd4bb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D -12.0 to -14.0 deg (C=11, 6mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR