4-(Trifluoromethoxy)phenyl isocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Isocyanato-4-(trifluoromethoxy)benzene
T2487-25G
T2487-5G
TCIAT2487-25GEA
349
EUR
TCIAT2487-25G
TCIAT2487-5G
4-(Trifluoromethoxy)phenyl isocyanate ≥98.0% (by GC)
4-(Trifluoromethoxy)phenyl isocyanate
Formule:
C₈H₄F₃NO₂ MW: 203.12 g/mol Kookpunt: 78°C Dichtheid: 1.34 Vlampunt: 69°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00035702 CAS nummer: 35037-73-1 EINECS: 252-328-0 UN: 2206 ADR: 6.1 III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...