4-Ethoxyphenyl isocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
E0770-25G
E0770-5G
TCIAE0770-25GEA
203
EUR
TCIAE0770-25G
TCIAE0770-5G
4-Ethoxyphenyl isocyanate ≥98.0% (by GC)
4-Ethoxyphenyl isocyanate
Formule:
C₉H₉NO₂ Kookpunt: 85°C Dichtheid: 1,11 g/cm³ (20 °C) Vlampunt: 106°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00013877 CAS nummer: 32459-62-4 UN: 2206 ADR: 6.1,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |