m-Anisidine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A0485-25G A0485-500G A0485-100G
TCIAA0485-25GEA 39.6 EUR
TCIAA0485-25G TCIAA0485-500G TCIAA0485-100G
m-Anisidine ≥98.0% (by GC, titration analysis)
m-Anisidine
Formule: C₇H₉NO
Kookpunt: 251°C
Smeltpunt: –2 °C
Dichtheid: 1,1 g/cm³ (20 °C)
Vlampunt: 126°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007783
CAS nummer: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1,III
Merck Index: 14,00667
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2353a723fe29f691866476bd6c750e5cb3b698e6&vid=2353a723fe29f691866476bd6c750e5cb3b698e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR