3-Aminothiophenol ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 3-Aminophenyl mercaptan, 3-Mercaptoaniline, 3-Aminobenzenethiol

A1852-25G A1852-5G
TCIAA1852-25GEA 245 EUR
TCIAA1852-25G TCIAA1852-5G
3-Aminothiophenol ≥97.0% (by GC, titration analysis)
3-Aminothiophenol
Formule: C₆H₇NS
Kookpunt: 190°C
Dichtheid: 1,1859 g/cm³ (20 °C)
Vlampunt: 79°C
MDL Nummer: MFCD00007791
CAS nummer: 22948-02-3
EINECS: 245-344-4
UN: 3267
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc29f9fe199def40cd3fb4631e48c4d3b95be889&vid=fc29f9fe199def40cd3fb4631e48c4d3b95be889&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR