6-Nitroindoline ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2,3-Dihydro-6-nitro-(1H)-indole
N0403-1G
N0403-5G
TCIAN0403-1GEA
17.6
EUR
TCIAN0403-1G
TCIAN0403-5G
6-Nitroindoline ≥97.0% (by GC, titration analysis)
6-Nitroindoline
Formule:
C₈H₈N₂O₂ Smeltpunt: 66…67 °C |
MDL Nummer:
MFCD00005710 CAS nummer: 19727-83-4 EINECS: 243-257-6 |
|
Specification Test Results
Appearance | Orange to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 67.0 to 70.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...