3-Nitrobenzyl alcohol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (3-Nitrophenyl)methanol

N0178-25G N0178-100G
TCIAN0178-25GEA 56.1 EUR
TCIAN0178-25G TCIAN0178-100G
3-Nitrobenzyl alcohol ≥97.0% (by GC)
3-Nitrobenzyl alcohol
Formule: C₇H₇NO₃
Kookpunt: 317 °C (1013 hPa)
Smeltpunt: 26…27 °C
Dichtheid: 1,3 g/cm³ (20 °C)
MDL Nummer: MFCD00007273
CAS nummer: 619-25-0
EINECS: 210-588-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=510a6c6df49055c6896003faa78e05503faceafb&vid=510a6c6df49055c6896003faa78e05503faceafb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR