5-Methoxyindole ≥99.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5-Methoxy-1H-indole, Indol-5-yl methyl ether
M0731-1G M0731-25G M0731-5G
TCIAM0731-1GEA 26.4 EUR
TCIAM0731-1G TCIAM0731-25G TCIAM0731-5G
5-Methoxyindole ≥99.0% (by GC)
5-Methoxyindole
Formule: C₉H₉NO
Kookpunt: 178°C
Smeltpunt: 55…56 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005674
CAS nummer: 1006-94-6
EINECS: 213-745-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1de83d98b8cd13dbbdbe625b55d102521ea3555&vid=f1de83d98b8cd13dbbdbe625b55d102521ea3555&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Brown powder to crystal
Purity (GC) min. 99.0 %
Melting point 54.0 to 58.0 deg-C
Solubility in Methanol very faint turbidity