Butyric anhydride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B0766-25ML B0766-500ML
TCIAB0766-25MLEA 17.6 EUR
TCIAB0766-25ML TCIAB0766-500ML
Butyric anhydride ≥98.0% (by titrimetric analysis)
Butyric anhydride
Formule: C₈H₁₄O₃
MW: 158.2 g/mol
Kookpunt: 198
Smeltpunt: -73
Vlampunt: 90°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009389
CAS nummer: 106-31-0
EINECS: 203-383-4
UN: 2739
ADR: 8 III
Merck Index: 14,01594
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b096153f9ac4c2fc704d91e39c8aebbcfe3b4696&vid=b096153f9ac4c2fc704d91e39c8aebbcfe3b4696&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (Morpholine Method) min. 98.0 %