Cyclohexylméthanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C0595-25G C0595-500G C0595-100G
TCIAC0595-25GEA 20.9 EUR
TCIAC0595-25G TCIAC0595-500G TCIAC0595-100G
Cyclohexylméthanol ≥98.0% (by GC)
Cyclohexylméthanol
Formule: C₇H₁₄O
Kookpunt: 183°C
Smeltpunt: –43 °C
Dichtheid: 0,926 g/cm³ (20 °C)
Vlampunt: 71°C
MDL Nummer: MFCD00001510
CAS nummer: 100-49-2
EINECS: 202-857-8
Merck Index: 14,02731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12549f15311b2e286e1825a6bc99692c63f042e7&vid=12549f15311b2e286e1825a6bc99692c63f042e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR