Trityl alcohol ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Triphenylmethanol, Triphenylcarbinol

T0516-25G T0516-500G
TCIAT0516-25GEA 17.6 EUR
TCIAT0516-25G TCIAT0516-500G
Trityl alcohol ≥99.0% (by GC)
Trityl alcohol
Formule: C₁₉H₁₆O
MW: 260.34 g/mol
Kookpunt: 380°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004445
CAS nummer: 76-84-6
EINECS: 200-988-5
Merck Index: 14,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bee50f3e3cc70047b0d6e366846c92361e253370&vid=bee50f3e3cc70047b0d6e366846c92361e253370&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 161.0 to 165.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR