3-Pyridone ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H0331-25G
H0331-500G
H0331-100G
TCIAH0331-25GEA
31.9
EUR
TCIAH0331-25G
TCIAH0331-500G
TCIAH0331-100G
3-Pyridone ≥98.0% (by GC, titration analysis)
3-Pyridone
Formule:
C₅H₅NO Kookpunt: 319 °C (1013 hPa) Smeltpunt: 124…126 °C Dichtheid: 1,2 g/cm³ (-70 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006378 CAS nummer: 109-00-2 EINECS: 203-637-4 |
|
Specification Test Results
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Solubility in hot Water | very faint turbidity |