Methaansulfonzuur ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

M0093-25G M0093-500G
TCIAM0093-25GEA 17.6 EUR
TCIAM0093-25G TCIAM0093-500G
Methaansulfonzuur ≥99.0% (by titrimetric analysis)
Methaansulfonzuur
Formule: CH₃SO₃H
Kookpunt: 122°C
Smeltpunt: 20 °C
Dichtheid: 1,48 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007518
CAS nummer: 75-75-2
EINECS: 200-898-6
UN: 2586
ADR: 8,III
Merck Index: 14,05954
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=914d103496c0b4dfb843b2649386213973912593&vid=914d103496c0b4dfb843b2649386213973912593&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR