Diphenyl sulphide ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Diphenylsulphane
P0230-25G P0230-500G
TCIAP0230-25GEA 18.7 EUR
TCIAP0230-25G TCIAP0230-500G
Diphenyl sulphide ≥98.0% (by GC)
Diphenyl sulphide
Formule: C₁₂H₁₀S
MW: 186.28 g/mol
Kookpunt: 296°C
Smeltpunt: –40°C
Vlampunt: 126°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003064
CAS nummer: 139-66-2
EINECS: 205-371-4
UN: 3082
ADR: 9 III
Merck Index: 14,07314
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6d99e2f0b92b30bcd5c7da8fc3cb5a586b3f4ea&vid=c6d99e2f0b92b30bcd5c7da8fc3cb5a586b3f4ea&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %