Hexamethyldisilane 98,0 GC_ASSAY_METHOD
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
H0638-10ML H0638-100ML
TCIAH0638-10MLEA 27.5 EUR
TCIAH0638-10ML TCIAH0638-100ML
Hexamethyldisilane 98,0 GC_ASSAY_METHOD
Hexamethyldisilane
Formule: C₆H₁₈Si₂
MW: 146.38 g/mol
Kookpunt: 113°C
Vlampunt: –9°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008258
CAS nummer: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44f58d1121bad72aef71c0da180e07dcfff3e856&vid=44f58d1121bad72aef71c0da180e07dcfff3e856&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %