2,3-Diethyl-5-methylpyrazine ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D1714-25ML D1714-5ML
TCIAD1714-25MLEA 154 EUR
TCIAD1714-25ML TCIAD1714-5ML
2,3-Diethyl-5-methylpyrazine ≥97.0% (by GC)
2,3-Diethyl-5-methylpyrazine
Formule: C₉H₁₄N₂
Kookpunt: 80°C
Dichtheid: 0,95 g/cm³ (20 °C)
Vlampunt: 57°C
MDL Nummer: MFCD00009635
CAS nummer: 18138-04-0
EINECS: 242-024-6
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57784f0d7fe63e7146a7c8a702c4d646aa915c60&vid=57784f0d7fe63e7146a7c8a702c4d646aa915c60&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %