Benzhydryl alcohol ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Diphenylcarbinol, Benzhydrol, Diphenylmethanol

D0898-25G D0898-500G
TCIAD0898-25GEA 17.6 EUR
TCIAD0898-25G TCIAD0898-500G
Benzhydryl alcohol ≥99.0% (by GC)
Benzhydryl alcohol
Formule: C₁₃H₁₂O
MW: 184.24 g/mol
Kookpunt: 297
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004488
CAS nummer: 91-01-0
EINECS: 202-033-8
Merck Index: 14,01090
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d538f1edeb64975b5552258e8aee5a7c3b864b4d&vid=d538f1edeb64975b5552258e8aee5a7c3b864b4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 66.0 to 68.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR