Cyclopropylméthanol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Cyclopropyl carbinol, Cyclopropanemethanol
C0982-25ML
C0982-5ML
TCIAC0982-25MLEA
60.5
EUR
TCIAC0982-25ML
TCIAC0982-5ML
Cyclopropylméthanol ≥98.0% (by GC)
Cyclopropylméthanol
Formule:
C₄H₈O Kookpunt: 123°C Smeltpunt: –60 °C Dichtheid: 0,904 g/cm³ (20 °C) Vlampunt: 35°C |
MDL Nummer:
MFCD00001309 CAS nummer: 2516-33-8 EINECS: 219-735-5 UN: 1987 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...