(R)-(+)-2-Bromopropionic acid ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B1756-25G B1756-5G
TCIAB1756-25GEA 291 EUR
TCIAB1756-25G TCIAB1756-5G
(R)-(+)-2-Bromopropionic acid ≥98.0% (by GC)
(R)-(+)-2-Bromopropionic acid
Formule: H₃CBrCHCOOH
Kookpunt: 104°C
Smeltpunt: 25 °C
Dichtheid: 1,693 g/cm³ (25 °C)
Vlampunt: 100°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00145205
CAS nummer: 10009-70-8
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f4e1b6ed5f73b8454defde9acccd775c48b190e&vid=2f4e1b6ed5f73b8454defde9acccd775c48b190e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 95.0 ee%
Specificaties
Environmentally Preferable