(R)-(-)-2-Octanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (R)-(-)-2-Hydroxy octane, (R)-2-Octanol, L(-)-2-Octanol, (R)-(-)-Hexyl methyl carbinol, (R)-(-)-Methyl hexyl carbinol

O0145-25ML O0145-5ML
TCIAO0145-25MLEA 486 EUR
TCIAO0145-25ML TCIAO0145-5ML
(R)-(-)-2-Octanol ≥98.0% (by GC)
(R)-(-)-2-Octanol
Formule: C₈H₁₈O
Kookpunt: 175 °C (1013 hPa)
Dichtheid: 0,8201 g/cm³ (20 °C)
Vlampunt: 71°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064284
CAS nummer: 5978-70-1
EINECS: 227-777-0
ADR: 3,III
Merck Index: 14,06752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b039b1907282caf2890e356ad97ed538b6f9a4b9&vid=b039b1907282caf2890e356ad97ed538b6f9a4b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -8.0 to -10.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR