(S)-(+)-Hydratropic acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P1220-1G
P1220-5G
TCIAP1220-1GEA
90.2
EUR
TCIAP1220-1G
TCIAP1220-5G
(S)-(+)-Hydratropic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-Hydratropic acid
Formule:
C₉H₁₀O₂ Kookpunt: 257 °C (1013 hPa) Smeltpunt: 30…31 °C Dichtheid: 1,1 g/cm³ (20 °C) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00063139 CAS nummer: 7782-24-3 |
|
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...