3,4-Difluorobenzyl alcohol ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (3,4-Difluorophenyl)methanol
D2415-25G D2415-5G
TCIAD2415-25GEA 151 EUR
TCIAD2415-25G TCIAD2415-5G
3,4-Difluorobenzyl alcohol ≥98.0% (by GC)
3,4-Difluorobenzyl alcohol
Formule: C₇H₆F₂O
Kookpunt: 197 °C (1013 hPa)
Smeltpunt: 9 °C
Dichtheid: 1,26 g/cm³ (20 °C)
Vlampunt: 97°C
MDL Nummer: MFCD00010628
CAS nummer: 85118-05-4
EINECS: 285-657-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0525e57b907b27157bc6cff291061157b96e354&vid=e0525e57b907b27157bc6cff291061157b96e354&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %