α,α'-Dichloro-m-xylene ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D1015-25G D1015-500G D1015-100G
TCIAD1015-25GEA 51.7 EUR
TCIAD1015-25G TCIAD1015-500G TCIAD1015-100G
α,α'-Dichloro-m-xylene ≥96.0% (by GC)
α,α'-Dichloro-m-xylene
Formule: C₈H₈Cl₂
Kookpunt: 254 °C (1013 hPa)
Smeltpunt: 33…35 °C
Dichtheid: 1,302 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000912
CAS nummer: 626-16-4
EINECS: 210-932-1
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb3f77977a182502111cfc38e3f0c23834f8b4d4&vid=fb3f77977a182502111cfc38e3f0c23834f8b4d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR