3,5-Dimethylphenyl isocyanate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D3474-25G D3474-5G
TCIAD3474-25GEA 163 EUR
TCIAD3474-25G TCIAD3474-5G
3,5-Dimethylphenyl isocyanate ≥98.0% (by GC)
3,5-Dimethylphenyl isocyanate
Formule: C₉H₉NO
Kookpunt: 205°C
Dichtheid: 1,04 g/cm³ (20 °C)
Vlampunt: 84°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00013868
CAS nummer: 54132-75-1
EINECS: 258-987-0
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4101c5760f270a0c3d7655df617a2fa9e048009e&vid=4101c5760f270a0c3d7655df617a2fa9e048009e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %