Hydrochinon ≥99.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1,4-Dihydroxybenzene, Quinol

H0186-25G H0186-500G
TCIAH0186-25GEA 17.6 EUR
TCIAH0186-25G TCIAH0186-500G
Hydrochinon ≥99.0% (by titrimetric analysis)
Hydrochinon
Formule: C₆H₄(OH)₂
MW: 110.11 g/mol
Kookpunt: 282°C
Smeltpunt: 172
Vlampunt: 165
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002339
CAS nummer: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9 III
Merck Index: 14,04808
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1617bcf6caad784bc4042b9b3ab9740f32ad8544&vid=1617bcf6caad784bc4042b9b3ab9740f32ad8544&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Redox method) min. 99.0 %
Melting point 172.0 to 176.0 deg-C
Solubility in dilute Acetic acid transparency
Ignition residue (Sulfate) max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR