1,4-Dihydroxynaphthalene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Naphthalene-1,4-diol, 1,4-Naphthalenediol, alpha-Naphthohydroquinone

D0589-25G D0589-5G
TCIAD0589-25GEA 289 EUR
TCIAD0589-25G TCIAD0589-5G
1,4-Dihydroxynaphthalene ≥98.0% (by GC)
1,4-Dihydroxynaphthalene
Formule: C₁₀H₈O₂
Smeltpunt: 191…193 °C
Dichtheid: 1,1 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003977
CAS nummer: 571-60-8
EINECS: 209-336-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27fa6a6697ac4ccf793a43528f23eb15bbb3bed2&vid=27fa6a6697ac4ccf793a43528f23eb15bbb3bed2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR