1,5-Decadiyne ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D1724-1ML D1724-5ML
TCIAD1724-1MLEA 68.2 EUR
TCIAD1724-1ML TCIAD1724-5ML
1,5-Decadiyne ≥97.0% (by GC)
1,5-Decadiyne
Formule: C₁₀H₁₄
Kookpunt: 79°C
MDL Nummer: MFCD00041651
CAS nummer: 53963-03-4
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50d3e1178df7cf5059cacf30797253cad00ed66c&vid=50d3e1178df7cf5059cacf30797253cad00ed66c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %