Diethyl glutarate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Glutaric acid diethyl ester, 1,5-Pentanedioic acid diethyl ester
G0184-25G
G0184-500G
TCIAG0184-25GEA
20.9
EUR
TCIAG0184-25G
TCIAG0184-500G
Diethyl glutarate ≥98.0% (by GC)
Diethyl glutarate
Formule:
C₉H₁₆O₄ Kookpunt: 230°C Vlampunt: 96°C |
MDL Nummer:
MFCD00009212 CAS nummer: 818-38-2 EINECS: 212-451-2 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...