1,7-Octadiyne ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

O0147-25ML O0147-5ML
TCIAO0147-25MLEA 145 EUR
TCIAO0147-25ML TCIAO0147-5ML
1,7-Octadiyne ≥98.0% (by GC)
1,7-Octadiyne
Formule: C₈H₁₀
Kookpunt: 135°C
Smeltpunt: –32 °C
Dichtheid: 0,8169 g/cm³ (21 °C)
Vlampunt: 23°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00008580
CAS nummer: 871-84-1
EINECS: 212-815-0
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e3590ab634c3167fe6cd25667f50a89469cdbe5&vid=1e3590ab634c3167fe6cd25667f50a89469cdbe5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR