Azelaic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid

A1318-25G A1318-250G
TCIAA1318-25GEA 25.3 EUR
TCIAA1318-25G TCIAA1318-250G
Azelaic acid ≥98.0% (by GC, titration analysis)
Azelaic acid
Formule: C₉H₁₆O₄
MW: 188.22 g/mol
Kookpunt: 306°C
Smeltpunt: 106
Vlampunt: 215
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004432
CAS nummer: 123-99-9
EINECS: 204-669-1
Merck Index: 14,00905
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=431eb9979a797a32adac0ea5ec6b0cfa3cac149b&vid=431eb9979a797a32adac0ea5ec6b0cfa3cac149b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 106.0 to 109.0 deg-C
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR