3,4-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D3373-1G D3373-5G
TCIAD3373-1GEA 37.4 EUR
TCIAD3373-1G TCIAD3373-5G
3,4-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
3,4-Difluorobenzylamine
Formule: C₇H₇F₂N
Kookpunt: 174 °C (1013 hPa)
Smeltpunt: 31 °C
Dichtheid: 1,21 g/cm³ (20 °C)
Vlampunt: 79°C
MDL Nummer: MFCD00010145
CAS nummer: 72235-53-1
EINECS: 276-503-6
UN: 3267
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ddf88e02b1fb633020bf1f8c5fc601ddbb081a6&vid=6ddf88e02b1fb633020bf1f8c5fc601ddbb081a6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %