4-Fluorobenzylamine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: (4-Fluorophenyl)methanamine
F0252-25G F0252-5G
TCIAF0252-25GEA 55 EUR
TCIAF0252-25G TCIAF0252-5G
4-Fluorobenzylamine ≥98.0% (by GC)
4-Fluorobenzylamine
Formule: C₇H₈FN
MW: 125.15 g/mol
Kookpunt: 183°C
Dichtheid: 1.09
Vlampunt: 66°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008120
CAS nummer: 140-75-0
EINECS: 205-430-4
UN: 2735
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36c5394a5f7550ad173a7ec3698371e5a44460db&vid=36c5394a5f7550ad173a7ec3698371e5a44460db&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %